Learn More
Meso Scale Discovery U-PLEX Human IL-3 ASY384-well
SDP
Catalog No. 5000080614
Encompass
The Meso Scale Discovery human IL-3 Singleplex Kit is an immunoassay that quantitatively measures human interleukin 3 (IL-3) in cell culture supernatants serum plasma and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay takes 3 5 hours to complete and has a sensitivity of 0 3 pg/mL
Catalog No. 50-000-80614 Supplier Meso Scale Discovery Supplier No. K251XMK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery human IL-3 Singleplex Kit is an immunoassay that quantitatively measures human interleukin 3 (IL-3) in cell culture supernatants serum plasma and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay takes 3 5 hours to complete and has a sensitivity of 0 3 pg/mL