Learn More
Meso Scale Discovery U-PLEX Human IL-31 ASY
SDP
Catalog No. 5000081859
Encompass
The Meso Scale Discovery U-PLEX Human IL-31 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-31 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The kit has a detection range of 1 53-2500 pg/mL and can quantify IL-31 levels in approximately 3 5 hours
Catalog No. 50-000-81859 Supplier Meso Scale Discovery Supplier No. K151WEK2
The Meso Scale Discovery U-PLEX Human IL-31 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-31 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The kit has a detection range of 1 53-2500 pg/mL and can quantify IL-31 levels in approximately 3 5 hours