Learn More
Meso Scale Discovery U-PLEX Human IL-31 ASY 25 PL
SDP
Catalog No. 5000082367
Encompass
The Meso Scale Discovery U-PLEX Human IL-31 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-31 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a diluent The assay has high specificity and shows no cross-reactivity with other cytokines The kit can perform up to 25 tests in a single run with results in 3 5 hours
Catalog No. 50-000-82367 Supplier Meso Scale Discovery Supplier No. K151WEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-31 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-31 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a diluent The assay has high specificity and shows no cross-reactivity with other cytokines The kit can perform up to 25 tests in a single run with results in 3 5 hours