Learn More
Meso Scale Discovery U-PLEX Human IL-31 ASY384-well
SDP
Catalog No. 5000081036
Encompass
The Meso Scale Discovery U-PLEX Human IL-31 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 31 (IL-31) in cell culture supernatants serum plasma (EDTA heparin) and saliva It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid ultra-sensitive and cost-effective multiplex immunoassay The kit includes a 384-well plate coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81036 Supplier Meso Scale Discovery Supplier No. K251WEK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-31 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 31 (IL-31) in cell culture supernatants serum plasma (EDTA heparin) and saliva It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid ultra-sensitive and cost-effective multiplex immunoassay The kit includes a 384-well plate coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL