Learn More
Meso Scale Discovery U-PLEX Human IL-33 ASY 25 PL
SDP
Catalog No. 5000081832
Encompass
The Meso Scale Discovery U-PLEX Human IL-33 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-33 with a lower limit of detection of 0 05 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator Blend The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81832 Supplier Meso Scale Discovery Supplier No. K151WFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-33 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-33 with a lower limit of detection of 0 05 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator Blend The assay takes approximately 3 5 hours to complete