Learn More
Meso Scale Discovery U-PLEX Human IL-33 ASY
SDP
Catalog No. 5000081422
Encompass
The Meso Scale Discovery U-PLEX Human IL-33 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-33 in human serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with Discovery Workbench software Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL
Catalog No. 50-000-81422 Supplier Meso Scale Discovery Supplier No. K151WFK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human IL-33 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-33 in human serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with Discovery Workbench software Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL