Learn More
Meso Scale Discovery U-PLEX Human IL-33 ASY
SDP
Catalog No. 5000081423
Encompass
The Meso Scale Discovery U-PLEX Human IL-33 Assay Kit is a multiplex immunoassay that measures IL-33 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized U-PLEX calibrant The assay requires 2 5 hours to complete and can test up to 80 samples in duplicate
Catalog No. 50-000-81423 Supplier Meso Scale Discovery Supplier No. K151WFK2
The Meso Scale Discovery U-PLEX Human IL-33 Assay Kit is a multiplex immunoassay that measures IL-33 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized U-PLEX calibrant The assay requires 2 5 hours to complete and can test up to 80 samples in duplicate