Learn More
Meso Scale Discovery U-PLEX Human IL-33 ASY384-well
SDP
Catalog No. 5000080979
Encompass
The Meso Scale Discovery U-PLEX Human IL-33 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-80979 Supplier Meso Scale Discovery Supplier No. K251WFK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-33 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL