Learn More
Meso Scale Discovery U-PLEX Human IL-4 ASY 25 PL
SDP
Catalog No. 5000079081
Encompass
The Meso Scale Discovery U-PLEX Human IL-4 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 4 (IL-4) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-4 with a lower limit of detection of 0 07 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The assay can be run on MSD s SECTOR Imager 6000 for separation-free assays
Catalog No. 50-000-79081 Supplier Meso Scale Discovery Supplier No. K151TWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-4 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 4 (IL-4) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-4 with a lower limit of detection of 0 07 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The assay can be run on MSD s SECTOR Imager 6000 for separation-free assays