Learn More
Meso Scale Discovery U-PLEX Human IL-4 ASY
SDP
Catalog No. 5000078520
Encompass
The Meso Scale Discovery U-PLEX Human IL-4 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 4 (IL-4) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary SECTOR Imager and QuickPlex SQ 120 instruments and provides high sensitivity a wide dynamic range and requires low sample volume The lower limit of detection is 0 07 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling
Catalog No. 50-000-78520 Supplier Meso Scale Discovery Supplier No. K151TWK1
The Meso Scale Discovery U-PLEX Human IL-4 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 4 (IL-4) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary SECTOR Imager and QuickPlex SQ 120 instruments and provides high sensitivity a wide dynamic range and requires low sample volume The lower limit of detection is 0 07 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling