Learn More
Meso Scale Discovery U-PLEX Human IL-4 ASY384-well
SDP
Catalog No. 5000080684
Encompass
The Meso Scale Discovery U-PLEX Human IL-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation with a detection limit of 0 04 pg/mL and a quantification limit of 1500 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium read buffer calibrator and diluent and is compatible with MSD s SECTOR Imager systems for high-throughput analysis
Catalog No. 50-000-80684 Supplier Meso Scale Discovery Supplier No. K251TWK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation with a detection limit of 0 04 pg/mL and a quantification limit of 1500 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium read buffer calibrator and diluent and is compatible with MSD s SECTOR Imager systems for high-throughput analysis