Learn More
Meso Scale Discovery U-PLEX Human IL-5 ASY 25 PL
SDP
Catalog No. 5000079145
Encompass
The Meso Scale Discovery U-PLEX Human IL-5 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and an IL-5 calibrator The assay has a lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-79145 Supplier Meso Scale Discovery Supplier No. K151UOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-5 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and an IL-5 calibrator The assay has a lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL