Learn More
Meso Scale Discovery U-PLEX Human IL-5 ASY
SDP
Catalog No. 5000079039
Encompass
The Meso Scale Discovery U-PLEX Human IL-5 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-5 with a detection range of 0 05-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator This assay allows for the accurate quantification of IL-5 which plays a role in eosinophil activation
Catalog No. 50-000-79039 Supplier Meso Scale Discovery Supplier No. K151UOK1
The Meso Scale Discovery U-PLEX Human IL-5 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-5 with a detection range of 0 05-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator This assay allows for the accurate quantification of IL-5 which plays a role in eosinophil activation