Learn More
Meso Scale Discovery U-PLEX Human IL-6 Assay, 384-well format, 0.33-2000 pg/mL dynamic range, Single-analyte detection, For research use only, Storage 2-8 °C, Kit size: 5 plates
SDP
Catalog No. NC3199586
Encompass
The MESO SCALE DISCOVERY U-PLEX Human IL-6 384-Well Assay is a single-analyte immunoassay specifically developed for measuring human Interleukin-6 (IL-6) in biological samples. This assay utilizes the U-PLEX platform, ensuring reliable and high-quality results while being optimized for 384-well plates. Its design allows for high sensitivity and a broad dynamic range, making it an excellent choice for various research applications.
  • Single-analyte detection for human IL-6.
  • Utilizes validated components for reliable results.
  • Compatible with plasma, serum, and cell culture supernatant.
  • Achieves a low limit of detection for subtle IL-6 level changes.
  • Dynamic range from 0.33 to 2,000 pg/mL.
  • Research Use Only statement.
  • Stored at 2-8 °C.
Catalog No. NC3199586 Supplier Meso Scale Discovery Supplier No. K251TXK2
Add to Cart
Add to Cart
Request a Quote
The MESO SCALE DISCOVERY U-PLEX Human IL-6 384-Well Assay is a single-analyte immunoassay specifically developed for measuring human Interleukin-6 (IL-6) in biological samples. This assay utilizes the U-PLEX platform, ensuring reliable and high-quality results while being optimized for 384-well plates. Its design allows for high sensitivity and a broad dynamic range, making it an excellent choice for various research applications.
  • Single-analyte detection for human IL-6.
  • Utilizes validated components for reliable results.
  • Compatible with plasma, serum, and cell culture supernatant.
  • Achieves a low limit of detection for subtle IL-6 level changes.
  • Dynamic range from 0.33 to 2,000 pg/mL.
  • Research Use Only statement.
  • Stored at 2-8 °C.