Learn More
Meso Scale Discovery U-PLEX HUMAN IL-6 ASSAY, 5 plates, 96-well format, Sandwich immunoassay, IL-6 analyte, Human species reactivity, Sensitivity 0.33 pg/mL, Dynamic range 0.33-2000 pg/mL
SDP
Catalog No. NC2906216
Encompass
U-PLEX HUMAN IL-6 ASSAY, 5 plates, 96-well format, Sandwich immunoassay, IL-6 analyte, Human species reactivity, Sensitivity 0.33 pg/mL, Dynamic range 0.33-2000 pg/mL
Catalog No. NC2906216 Supplier Meso Scale Discovery Supplier No. K151TXK2
U-PLEX HUMAN IL-6 ASSAY, 5 plates, 96-well format, Sandwich immunoassay, IL-6 analyte, Human species reactivity, Sensitivity 0.33 pg/mL, Dynamic range 0.33-2000 pg/mL