Learn More
Meso Scale Discovery U-PLEX Human IL-6 ASYQP
SDP
Catalog No. 5000080502
Encompass
The U-PLEX Human IL-6 Assay (K151TXK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of interleukin-6 protein levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-6 quantification in research applications including inflammation studies biomarker discovery and cytokine profiling workflows
Catalog No. 50-000-80502 Supplier Meso Scale Discovery Supplier No. K151TXK22
The U-PLEX Human IL-6 Assay (K151TXK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of interleukin-6 protein levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-6 quantification in research applications including inflammation studies biomarker discovery and cytokine profiling workflows