Learn More
Meso Scale Discovery U-PLEX Human IL-7 ASY 25 PL
SDP
Catalog No. 5000079146
Encompass
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-7 with a detection range of 0 05-1000 pg/mL and a lower limit of detection of 0 03 pg/mL The assay can be used to monitor IL-7 levels in immune system studies involving T and B cell development bone loss and autoimmune diseases The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent
Catalog No. 50-000-79146 Supplier Meso Scale Discovery Supplier No. K151UPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-7 with a detection range of 0 05-1000 pg/mL and a lower limit of detection of 0 03 pg/mL The assay can be used to monitor IL-7 levels in immune system studies involving T and B cell development bone loss and autoimmune diseases The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent