Learn More
Meso Scale Discovery U-PLEX Human IL-7 ASY
SDP
Catalog No. 5000079040
Encompass
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 04 pg/mL and the assay measures IL-7 concentrations from 0 08 to 1000 pg/mL
Catalog No. 50-000-79040 Supplier Meso Scale Discovery Supplier No. K151UPK2
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 04 pg/mL and the assay measures IL-7 concentrations from 0 08 to 1000 pg/mL