Learn More
Meso Scale Discovery U-PLEX Human IL-7 ASY384-well
SDP
Catalog No. 5000080978
Encompass
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is designed for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants using an electrochemiluminescent detection technology The assay has high sensitivity and wide dynamic range It includes a plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The kit can perform up to 80 tests in a single microplate with a testing time of 3 5 hours
Catalog No. 50-000-80978 Supplier Meso Scale Discovery Supplier No. K251UPK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is designed for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants using an electrochemiluminescent detection technology The assay has high sensitivity and wide dynamic range It includes a plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The kit can perform up to 80 tests in a single microplate with a testing time of 3 5 hours