Learn More
Meso Scale Discovery U-PLEX Human IL-7 ASYQP
SDP
Catalog No. 5000080539
Encompass
The U-PLEX Human IL-7 Assay (K151UPK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-7 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-7 quantification in research applications
Catalog No. 50-000-80539 Supplier Meso Scale Discovery Supplier No. K151UPK22
The U-PLEX Human IL-7 Assay (K151UPK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-7 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-7 quantification in research applications