Learn More
Meso Scale Discovery U-PLEX Human IL-8 ASY
SDP
Catalog No. 5000080503
Encompass
The Meso Scale Discovery U-PLEX Human IL-8 Assay Kit is a multiplex immunoassay that measures IL-8 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80503 Supplier Meso Scale Discovery Supplier No. K151TYK1
The Meso Scale Discovery U-PLEX Human IL-8 Assay Kit is a multiplex immunoassay that measures IL-8 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL