Learn More
Meso Scale Discovery U-PLEX Human IL-8 ASYQP
SDP
Catalog No. 5000079673
Encompass
The U-PLEX Human IL-8 Assay (K151TYK) is a multiplex immunoassay platform from Meso Scale Discovery designed for quantitative measurement of interleukin-8 (IL-8) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers for comprehensive cytokine profiling in research applications
Catalog No. 50-000-79673 Supplier Meso Scale Discovery Supplier No. K151TYK21
The U-PLEX Human IL-8 Assay (K151TYK) is a multiplex immunoassay platform from Meso Scale Discovery designed for quantitative measurement of interleukin-8 (IL-8) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers for comprehensive cytokine profiling in research applications