Learn More
Meso Scale Discovery U-PLEX Human IL-9 ASY 25 PL
SDP
Catalog No. 5000081793
Encompass
The Meso Scale Discovery U-PLEX Human IL-9 Assay Kit is a multiplex immunoassay for measuring IL-9 in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and diluents Sensitivity is 0 3 pg/mL with an upper limit of 10 000 pg/mL It requires 50 L of sample and is compatible with MSD instruments offering specific and rapid IL-9 measurement
Catalog No. 50-000-81793 Supplier Meso Scale Discovery Supplier No. K151XKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-9 Assay Kit is a multiplex immunoassay for measuring IL-9 in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and diluents Sensitivity is 0 3 pg/mL with an upper limit of 10 000 pg/mL It requires 50 L of sample and is compatible with MSD instruments offering specific and rapid IL-9 measurement