Learn More
Meso Scale Discovery U-PLEX Human IL-9 ASY
SDP
Catalog No. 5000078753
Encompass
The Meso Scale Discovery U-PLEX Human IL-9 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-9 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide a sensitive and reproducible method for measuring IL-9 with a detection range of 0 05-1000 pg/mL and an LLOQ of 0 05 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator Blend 1 The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78753 Supplier Meso Scale Discovery Supplier No. K151XKK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human IL-9 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-9 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide a sensitive and reproducible method for measuring IL-9 with a detection range of 0 05-1000 pg/mL and an LLOQ of 0 05 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator Blend 1 The assay takes approximately 3 5 hours to complete