Learn More
Meso Scale Discovery U-PLEX Human IL-9 ASY384-well
SDP
Catalog No. 5000080973
Encompass
The Meso Scale Discovery U-PLEX Human IL-9 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 9 (IL-9) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label read buffer 10X wash buffer blocker solutions calibrator diluent and a vial of lyophilized calibrator The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range from 0 04-1500 pg/mL
Catalog No. 50-000-80973 Supplier Meso Scale Discovery Supplier No. K251XKK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-9 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 9 (IL-9) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label read buffer 10X wash buffer blocker solutions calibrator diluent and a vial of lyophilized calibrator The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range from 0 04-1500 pg/mL