Learn More
Meso Scale Discovery U-PLEX HUMAN IP-10 ASSAY, Single-analyte immunoassay, 96-well plate, Dynamic Range: 0.49-6,000 pg/mL, LLOD: 0.49 pg/mL
SDP
Supplier:  Meso Scale Discovery K151UFK2
Encompass
The U-PLEX Human IP-10 Assay is a single-analyte immunoassay designed for the sensitive detection of human IP-10 (CXCL10). Utilizing the U-PLEX platform, this assay ensures high-quality results across various sample types, such as serum, plasma, and cell culture supernatant. It is intended for research applications in immunology and inflammation, providing flexibility for diverse experimental designs.
  • Reliable quantification with validated components
  • Broad detection range from 0.49 to 6,000 pg/mL
  • High sensitivity, detecting low levels of IP-10
  • Compatible with multiple sample types
  • Research-grade for immunology and inflammation studies
Catalog No. NC1676324