Learn More
Meso Scale Discovery U-PLEX Human IP-10 ASY
SDP
Catalog No. 5000079038
Encompass
The Meso Scale Discovery U-PLEX Human IP-10 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity with no cross-reactivity and 1% interference It requires 50 L or less sample volume and has an ultra-low lower limit of detection of 45 fg/mL The assay takes 3 5 hours for completion This powerful and sensitive assay allows reliable quantification of IP-10 across a wide range of concentrations
Catalog No. 50-000-79038 Supplier Meso Scale Discovery Supplier No. K151UFK1
The Meso Scale Discovery U-PLEX Human IP-10 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity with no cross-reactivity and 1% interference It requires 50 L or less sample volume and has an ultra-low lower limit of detection of 45 fg/mL The assay takes 3 5 hours for completion This powerful and sensitive assay allows reliable quantification of IP-10 across a wide range of concentrations