Learn More
Meso Scale Discovery U-PLEX Human LAG3 ASY 25 PL
SDP
Catalog No. 5000078575
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is designed for the quantitative measurement of LAG3 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for detection of human LAG3 with no cross-reactivity to related molecules It includes a plate coated with capture antibody detection antibody and calibrator and control solutions The kit allows for up to 80 tests in a single run with results in 3 5 hours
Catalog No. 50-000-78575 Supplier Meso Scale Discovery Supplier No. K151Q7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is designed for the quantitative measurement of LAG3 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for detection of human LAG3 with no cross-reactivity to related molecules It includes a plate coated with capture antibody detection antibody and calibrator and control solutions The kit allows for up to 80 tests in a single run with results in 3 5 hours