Learn More
Meso Scale Discovery U-PLEX Human LAG3 ASY
SDP
Catalog No. 5000081263
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay that enables simultaneous detection and quantification of human LAG3 and other biomarkers of interest from a single sample This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve It is compatible with serum plasma and cell culture supernatant samples The U-PLEX assay delivers rapid results in just 3 5 hours
Catalog No. 50-000-81263 Supplier Meso Scale Discovery Supplier No. K151Q7K2
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay that enables simultaneous detection and quantification of human LAG3 and other biomarkers of interest from a single sample This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve It is compatible with serum plasma and cell culture supernatant samples The U-PLEX assay delivers rapid results in just 3 5 hours