Learn More
Meso Scale Discovery U-PLEX Human LAG3 ASY384-well
SDP
Catalog No. 5000081045
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 384-Well Assay is a multiplex immunoassay kit for quantifying human LAG3 protein in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption The kit includes a 384-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and controls It requires only 25 L per well with a detection limit of 45 pg/mL and an upper quantitation limit of 50 000 pg/mL for LAG3
Catalog No. 50-000-81045 Supplier Meso Scale Discovery Supplier No. K251Q7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human LAG3 384-Well Assay is a multiplex immunoassay kit for quantifying human LAG3 protein in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption The kit includes a 384-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and controls It requires only 25 L per well with a detection limit of 45 pg/mL and an upper quantitation limit of 50 000 pg/mL for LAG3