Learn More
Meso Scale Discovery U-PLEX Human LAG3 ASYQP
SDP
Catalog No. 5000078477
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay that enables simultaneous quantification of LAG3 and other human biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for customization and multiplexing with other U-PLEX biomarker kits for immune monitoring biomarker discovery and immunology research
Catalog No. 50-000-78477 Supplier Meso Scale Discovery Supplier No. K151Q7K22
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay that enables simultaneous quantification of LAG3 and other human biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for customization and multiplexing with other U-PLEX biomarker kits for immune monitoring biomarker discovery and immunology research