Learn More
Meso Scale Discovery U-PLEX Human LAG3 ASYQP
SDP
Catalog No. 5000082308
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay for detecting and quantifying human LAG3 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL for LAG3
Catalog No. 50-000-82308 Supplier Meso Scale Discovery Supplier No. K151Q7K21
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay for detecting and quantifying human LAG3 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL for LAG3