Learn More
Meso Scale Discovery U-PLEX Human LH ASY384-well
SDP
Catalog No. 5000081102
Encompass
The Meso Scale Discovery U-PLEX Human LH 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of luteinizing hormone (LH) in human serum plasma and culture media samples It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a broad dynamic range of 0 03-1000 mIU/mL and requires as little as 25 L of sample It includes a plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The streamlined no-wash protocol allows completion in just 3 5 hours
Catalog No. 50-000-81102 Supplier Meso Scale Discovery Supplier No. K2516FK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human LH 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of luteinizing hormone (LH) in human serum plasma and culture media samples It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a broad dynamic range of 0 03-1000 mIU/mL and requires as little as 25 L of sample It includes a plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The streamlined no-wash protocol allows completion in just 3 5 hours