Learn More
Meso Scale Discovery U-PLEX Human LIGHT/TNFSF14 ASY
SDP
Catalog No. 5000080144
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of TNF Superfamily Member 14 (TNFSF14) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and plate seals The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL for TNFSF14
Catalog No. 50-000-80144 Supplier Meso Scale Discovery Supplier No. K151AHCK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of TNF Superfamily Member 14 (TNFSF14) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and plate seals The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL for TNFSF14