Learn More
Meso Scale Discovery U-PLEX Human M-CSF ASY 25 PL
SDP
Catalog No. 5000079628
Encompass
The Meso Scale Discovery U-PLEX Human M-CSF Assay is a multiplex immunoassay kit for quantifying macrophage colony-stimulating factor (M-CSF) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The 30-plex kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It detects M-CSF with a lower limit of 45 4 pg/mL and an upper limit of 100 000 pg/mL
Catalog No. 50-000-79628 Supplier Meso Scale Discovery Supplier No. K151XRK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human M-CSF Assay is a multiplex immunoassay kit for quantifying macrophage colony-stimulating factor (M-CSF) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The 30-plex kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It detects M-CSF with a lower limit of 45 4 pg/mL and an upper limit of 100 000 pg/mL