Learn More
Meso Scale Discovery U-PLEX Human M-CSF ASY
SDP
Catalog No. 5000080090
Encompass
The Meso Scale Discovery U-PLEX Human M-CSF Assay is a multiplex immunoassay kit for measuring macrophage colony-stimulating factor (M-CSF) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The 30-plex kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls The workflow is completed in 3 5 hours with a detection limit of 45 pg/mL ideal for accurate M-CSF quantification
Catalog No. 50-000-80090 Supplier Meso Scale Discovery Supplier No. K151XRK2
The Meso Scale Discovery U-PLEX Human M-CSF Assay is a multiplex immunoassay kit for measuring macrophage colony-stimulating factor (M-CSF) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The 30-plex kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls The workflow is completed in 3 5 hours with a detection limit of 45 pg/mL ideal for accurate M-CSF quantification