Learn More
Meso Scale Discovery U-PLEX Human M-CSF ASY384-well
SDP
Catalog No. 5000081038
Encompass
The Meso Scale Discovery Human M-CSF Singleplex Assay Kit is a singleplex immunoassay that quantitatively measures M-CSF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator It can detect M-CSF concentrations between 0 3-10 000 pg/mL with an incubation time of 2 hours
Catalog No. 50-000-81038 Supplier Meso Scale Discovery Supplier No. K251XRK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human M-CSF Singleplex Assay Kit is a singleplex immunoassay that quantitatively measures M-CSF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator It can detect M-CSF concentrations between 0 3-10 000 pg/mL with an incubation time of 2 hours