Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery U-PLEX Human MCP-1 Assay, 384-well format, Single-analyte, Electrochemiluminescence (ECL), Research Use Only
SDP

Catalog No. NC2866720
Encompass

The MESO SCALE DISCOVERY U-PLEX Human MCP-1 Assay is an advanced electrochemiluminescence (ECL) immunoassay designed for the accurate quantitative detection of human Monocyte Chemoattractant Protein-1 (MCP-1) in a 384-well plate format. This assay is part of the versatile U-PLEX platform, which allows for single-analyte measurements and the capability of multiplexing for extensive biomarker analysis across various sample types, including serum and plasma. Intended for research use only, this product ensures reliable findings in scientific investigations.

  • Analyte: Human MCP-1
  • Plate format: 384-well
  • Assay type: Single-analyte (can be multiplexed)
  • Detection technology: Electrochemiluminescence (ECL)
  • Research use only: true
  • Product status: Made-to-order
  • Minimum order size: 25 plates
  • Compatible sample types: Serum, Plasma
  • Included diluents: Diluent 3, Diluent 57

Catalog No. NC2866720 Supplier Meso Scale Discovery Supplier No. K251UGK4
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The MESO SCALE DISCOVERY U-PLEX Human MCP-1 Assay is an advanced electrochemiluminescence (ECL) immunoassay designed for the accurate quantitative detection of human Monocyte Chemoattractant Protein-1 (MCP-1) in a 384-well plate format. This assay is part of the versatile U-PLEX platform, which allows for single-analyte measurements and the capability of multiplexing for extensive biomarker analysis across various sample types, including serum and plasma. Intended for research use only, this product ensures reliable findings in scientific investigations.

  • Analyte: Human MCP-1
  • Plate format: 384-well
  • Assay type: Single-analyte (can be multiplexed)
  • Detection technology: Electrochemiluminescence (ECL)
  • Research use only: true
  • Product status: Made-to-order
  • Minimum order size: 25 plates
  • Compatible sample types: Serum, Plasma
  • Included diluents: Diluent 3, Diluent 57

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.