Learn More
Meso Scale Discovery U-PLEX Human MCP-1 ASY 25 PL
SDP
Catalog No. 5000080395
Encompass
The Meso Scale Discovery U-PLEX Human MCP-1 Assay Kit quantifies monocyte chemoattractant protein 1 (MCP-1) in human serum plasma and cell culture supernatants Using MSD s SULFO-TAG labels and multi-array technology it measures MCP-1 levels down to 0 4 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies buffers blockers calibrators and controls The assay takes about 3 5 hours and requires an MSD instrument like the MESO QuickPlex SQ 120 It offers high sensitivity broad range and minimal sample volume for efficient MCP-1 analysis
Catalog No. 50-000-80395 Supplier Meso Scale Discovery Supplier No. K151UGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MCP-1 Assay Kit quantifies monocyte chemoattractant protein 1 (MCP-1) in human serum plasma and cell culture supernatants Using MSD s SULFO-TAG labels and multi-array technology it measures MCP-1 levels down to 0 4 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies buffers blockers calibrators and controls The assay takes about 3 5 hours and requires an MSD instrument like the MESO QuickPlex SQ 120 It offers high sensitivity broad range and minimal sample volume for efficient MCP-1 analysis