Learn More
Meso Scale Discovery U-PLEX Human MCP-2 ASY 25 PL
SDP
Catalog No. 5000081813
Encompass
The Meso Scale Discovery U-PLEX Human MCP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human chemokine MCP-2 It utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies calibrators and buffers The assay has high sensitivity and wide dynamic range requires low sample volume and provides rapid results It is intended for research use only to monitor MCP-2 levels in cell culture supernatants serum plasma and other biological samples
Catalog No. 50-000-81813 Supplier Meso Scale Discovery Supplier No. K151XHK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MCP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human chemokine MCP-2 It utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies calibrators and buffers The assay has high sensitivity and wide dynamic range requires low sample volume and provides rapid results It is intended for research use only to monitor MCP-2 levels in cell culture supernatants serum plasma and other biological samples