Learn More
Meso Scale Discovery U-PLEX Human MCP-2 ASYQP
SDP
Catalog No. 5000079174
Encompass
The U-PLEX Human MCP-2 Assay (K151XHK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of human monocyte chemoattractant protein-2 (MCP-2/CCL8) in biological samples This electrochemiluminescence-based assay offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods The U-PLEX format allows for flexible scalable testing with excellent precision and reproducibility for research applications in inflammation immunology and biomarker studies
Catalog No. 50-000-79174 Supplier Meso Scale Discovery Supplier No. K151XHK22
The U-PLEX Human MCP-2 Assay (K151XHK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of human monocyte chemoattractant protein-2 (MCP-2/CCL8) in biological samples This electrochemiluminescence-based assay offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods The U-PLEX format allows for flexible scalable testing with excellent precision and reproducibility for research applications in inflammation immunology and biomarker studies