Learn More
Meso Scale Discovery U-PLEX Human MCP-3 ASY 25 PL
SDP
Catalog No. 5000080092
Encompass
The Meso Scale Discovery U-PLEX Human MCP-3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human MCP-3 biomarker in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents needed to perform the immunoassay It can be used for biomarker research and drug discovery applications related to inflammation autoimmune disorders and oncology
Catalog No. 50-000-80092 Supplier Meso Scale Discovery Supplier No. K151XIK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MCP-3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human MCP-3 biomarker in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents needed to perform the immunoassay It can be used for biomarker research and drug discovery applications related to inflammation autoimmune disorders and oncology