Learn More
Meso Scale Discovery U-PLEX Human MCP-3 ASY384-well
SDP
Catalog No. 5000080717
Encompass
The Meso Scale Discovery U-PLEX Human MCP-3 (384-well) Assay Kit is designed for quantifying human MCP-3 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a sensitive reproducible assay with low sample volume needs The kit includes a 384-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Sensitivity is 0 3 pg/mL with a quantitation limit of 10 000 pg/mL
Catalog No. 50-000-80717 Supplier Meso Scale Discovery Supplier No. K251XIK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MCP-3 (384-well) Assay Kit is designed for quantifying human MCP-3 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a sensitive reproducible assay with low sample volume needs The kit includes a 384-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Sensitivity is 0 3 pg/mL with a quantitation limit of 10 000 pg/mL