Learn More
Meso Scale Discovery U-PLEX Human MDC ASY 25 PL
SDP
Catalog No. 5000080376
Encompass
The U-PLEX Human MDC Assay K151UIK from Meso Scale Diagnostics is a multiplex immunoassay panel that measures 10 human inflammatory biomarkers from 50 L of serum plasma (citrate heparin) or cell culture supernatant This electrochemiluminescent assay quantitatively measures IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- with high sensitivity and dynamic range The kit includes a 96-well MULTI-ARRAY plate calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals sufficient for one 96-well plate This assay allows efficient and accurate measurement of multiple human inflammatory mediators from small sample volumes
Catalog No. 50-000-80376 Supplier Meso Scale Discovery Supplier No. K151UIK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MDC Assay K151UIK from Meso Scale Diagnostics is a multiplex immunoassay panel that measures 10 human inflammatory biomarkers from 50 L of serum plasma (citrate heparin) or cell culture supernatant This electrochemiluminescent assay quantitatively measures IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- with high sensitivity and dynamic range The kit includes a 96-well MULTI-ARRAY plate calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals sufficient for one 96-well plate This assay allows efficient and accurate measurement of multiple human inflammatory mediators from small sample volumes