Learn More
Meso Scale Discovery U-PLEX Human MDC ASY
SDP
Catalog No. 5000079678
Encompass
The U-PLEX Human MDC Assay K151UIK from Meso Scale Discovery is a multiplex immunoassay panel that measures 10 human inflammatory biomarkers This assay kit allows simultaneous quantitation of IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- in serum plasma and culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay requires an MSD instrument such as QuickPlex SECTOR Imager or SQ 120 for analysis With minimal sample volume requirements and rapid results this panel is ideal for inflammation and cytokine research
Catalog No. 50-000-79678 Supplier Meso Scale Discovery Supplier No. K151UIK2
The U-PLEX Human MDC Assay K151UIK from Meso Scale Discovery is a multiplex immunoassay panel that measures 10 human inflammatory biomarkers This assay kit allows simultaneous quantitation of IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- in serum plasma and culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay requires an MSD instrument such as QuickPlex SECTOR Imager or SQ 120 for analysis With minimal sample volume requirements and rapid results this panel is ideal for inflammation and cytokine research