Learn More
Meso Scale Discovery U-PLEX Human MDC ASY384-well
SDP
Catalog No. 5000080974
Encompass
The Meso Scale Discovery Human MDC Singleplex Assay measures Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection of MDC with a detection limit of 19 5 pg/mL and quantification up to 10 000 pg/mL It requires 50 L of sample per well and is read on MSD s SECTOR Imager systems providing a rapid accurate method for measuring MDC important for immune function
Catalog No. 50-000-80974 Supplier Meso Scale Discovery Supplier No. K251UIK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human MDC Singleplex Assay measures Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection of MDC with a detection limit of 19 5 pg/mL and quantification up to 10 000 pg/mL It requires 50 L of sample per well and is read on MSD s SECTOR Imager systems providing a rapid accurate method for measuring MDC important for immune function