Learn More
Meso Scale Discovery U-PLEX Human MDC ASYQP
SDP
Catalog No. 5000080535
Encompass
The U-PLEX Human MDC Assay (K151UIK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of macrophage-derived chemokine (MDC/CCL22) in biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and controls for reliable reproducible results in research applications studying inflammatory responses and immune cell recruitment
Catalog No. 50-000-80535 Supplier Meso Scale Discovery Supplier No. K151UIK22
The U-PLEX Human MDC Assay (K151UIK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of macrophage-derived chemokine (MDC/CCL22) in biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and controls for reliable reproducible results in research applications studying inflammatory responses and immune cell recruitment