Learn More
Meso Scale Discovery U-PLEX Human MDC ASYQP
SDP
Catalog No. 5000082023
Encompass
The U-PLEX Human MDC Assay (K151UIK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure macrophage-derived chemokine (MDC/CCL22) levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantitative detection of MDC a key chemokine involved in immune responses and inflammation The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications
Catalog No. 50-000-82023 Supplier Meso Scale Discovery Supplier No. K151UIK21
Add to Cart
Add to Cart
The U-PLEX Human MDC Assay (K151UIK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure macrophage-derived chemokine (MDC/CCL22) levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantitative detection of MDC a key chemokine involved in immune responses and inflammation The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications