Learn More
Meso Scale Discovery U-PLEX Human MIF ASY 25 PL
SDP
Catalog No. 5000080462
Encompass
The Meso Scale Discovery U-PLEX Human MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in human serum plasma urine saliva and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less sample volume and is compatible with the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and necessary reagents
Catalog No. 50-000-80462 Supplier Meso Scale Discovery Supplier No. K151XJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in human serum plasma urine saliva and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less sample volume and is compatible with the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and necessary reagents