Learn More
Meso Scale Discovery U-PLEX Human MIF ASY
SDP
Catalog No. 5000079213
Encompass
The Meso Scale Discovery U-PLEX Human MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in various human samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with necessary antibodies calibrators and controls It processes up to 80 samples in 3 5 hours aiding in the study of MIF s role in inflammatory and autoimmune diseases
Catalog No. 50-000-79213 Supplier Meso Scale Discovery Supplier No. K151XJK2
The Meso Scale Discovery U-PLEX Human MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in various human samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with necessary antibodies calibrators and controls It processes up to 80 samples in 3 5 hours aiding in the study of MIF s role in inflammatory and autoimmune diseases